Typical diet - trace elements, NIST-1548A
Lieferant: LGC Standards
PROONIST-1548AEA 0 EUR
PROONIST-1548A
Typical diet - trace elements, NIST-1548A
This standard reference material is intended to validate analytical methods for the determination of proximate content and minor and trace constituent elements in mixed diets and food matrices. Used for metabolic studies.
A unit consists of two bottles, each containing approximately 6.5 g of the freeze-dried homogenate of mixed diet foods.
Standards: Traceable to NIST and SRM numbers.