Chamazulen, ChromaDex

Lieferant: LGC Standards
PROOCDX00003383010EA 266 EUR
PROOCDX00003383010 PROOCDX00003383100
Chamazulen, ChromaDex
Standards Organische Standards
This chamazulen is a organic component which have been more commonly used as phytochemical standards. It has also been used for forensic and clinical analysis.

  • Liquid form
  • Blue in colour
  • Odourless in nature
  • Flash point: 4,0 °C
  • Boiling point: 298 °C
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR