Cool unit –10 °C for tissue embedding system, TES 99
Lieferant: Medite
MEDT02-9942-00EA 0 EUR
MEDT02-9942-00
Cool unit –10 °C for tissue embedding system, TES 99
Kühlbehälter Tischkühler
The compact model with small foot print for laboratories with limited table space.
This cool unit can accomodate up to 80 blocks that can be chilled at a temperature adjustable down to –10 °C.