Conductivity meter, bench, Eutech CON 700
Lieferant: Thermo Fisher Scientific
EUTEECCON70043SEA 1070 EUR
EUTEECCON70043S
Conductivity meter, bench, Eutech CON 700
This economical and user-friendly conductivity meter is ideal for use in laboratories, productions plants and schools.
  • Large screen displays conductivity or TDS readings with temperature in °C or °F
  • Ready indicator informs users when readings are stable
  • Selectable cell constant
  • Auto-ranging across five conductivity and TDS ranges
  • Up to 5-point push button calibration
  • Non volatile memory holds up to 100 data points
  • Integral electrode holder
Lieferumfang: Supplied with conductivity/ATC probe, integral electrode holder and 100/240 VAC adapter.