Filter tips, sterile, WHEATON®
Lieferant: DWK Life Sciences
WHEA851180-01EA 1140 EUR
WHEA851180-01 WHEA851180-02
Filter tips, sterile, WHEATON®
Pipettenspitzen
These filter tips are made from high grade, virgin, metal-free PP.
  • Precision injection molding care for air-tight fit
  • Lot-by-lot consistency
  • High-density filter prevents aerosol contamination
  • DNase and RNase-free
  • Pyrogen-free