Anti-SLIT2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB30933EA 545 EUR
ABNOPAB30933
Anti-SLIT2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against partial recombinant human SLIT2.

Type: Primary
Antigen: SLIT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR