Anti-Ly6g Rat Monoclonal Antibody (PerCP) [clone: 1A8]
Lieferant: Abnova
ABNOMAB15467EA 440 EUR
ABNOMAB15467
Anti-Ly6g Rat Monoclonal Antibody (PerCP) [clone: 1A8]
Antikörper
Rat monoclonal antibody raised against native mouse Ly6g.
Type: Primary
Antigen: Ly6g
Clonality: Monoclonal
Clone: 1A8
Conjugation: PerCP
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse