Plasmid isolation, RapidPURE™ Plasmid Mini kit
Lieferant: MP Biomedicals
RapidPURE™
ICNA112066200EA 131 EUR
ICNA112066200 ICNA112066400 ICNA112066600
Plasmid isolation, RapidPURE™ Plasmid Mini kit
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
The RapidPURE™ Plasmid Mini Kit is designed to isolate up to 20 μg of plasmid DNA from 1,5 ml of an overnight bacterial culture for use in routine molecular biology applications, such as fluorescent and radioactive sequencing and cloning.
The plasmid purification process employs a patented Turbo Filter containing a GLASSMILK®-embedded membrane. The Turbo Filter is combined with a catch tube into a Turbo Cartridge featuring one hole in the center of the cap for the in-situ addition of wash solution, and a second hole permitting in-situ removal of waste solution using vacuum aspiration.
Lieferumfang: The kit contains Pre-lysis buffer, alkaline lysis solution, neutralising solution, RapidPURE® mini salt solution, wash solution, RapidPURE® turbo cartridges, RapidPURE® Turbo recovery tubes and CIRCLEGROW® broth pouch(es).