Thermometer armour case, SP Bel-Art
Lieferant: Bel-Art Products, a part of SP
BELAB614009400EA 79.5 EUR
BELAB614009400
Thermometer armour case, SP Bel-Art
Thermometer-Halterungen
This nickel-plated brass case protects thermometers from accidental breakage during use and storage.
  • Screw cap
  • Ring for hanging
  • Lower section surrounding the bulb with small openings for circulation