Cell culture media, Ham's F10 (10X)
Lieferant: MP Biomedicals
ICNA091440049EA 30.2 EUR
ICNA091440049
Cell culture media, Ham's F10 (10X)
Zellkulturmedien
Ham's nutrient mixtures were developed for the clonal growth of several chinese hamster ovary (CHO) clone cells, Hela cells and mouse L-cells.
  • Ideal for mammalian cell culture
  • Available as clear, orange solution
  • pH of 5,5 to 6,1
  • To be stored at 4 °C
Ham's F-10 has demonstrated satisfactory growth of human diploid cells, white blood cells for chromosomal analysis and primary rat, rabbit and chicken tissue explants. Both F-10 and F-12 are formulated for use with or without serum, depending on the type of cells being cultured.