Cell culture media, high growth enhancement medium
Lieferant: MP Biomedicals
ICNA091033822EA 65.65 EUR
ICNA091033822
Cell culture media, high growth enhancement medium
Zellkulturmedien
High Growth Enhancement Medium is a modification of DMEM in which 3,6 g/L fructose is substituted for glucose, resulting in better pH control and higher cell yields. Fructose is metabolised slower than glucose, resulting in slower acid build up. This media is recommended for the growth and support of cells on microcarrier matrices.