Syringes, PICO50
Lieferant: Radiometer
RADC956-552EA 0 EUR
RADC956-552
Syringes, PICO50
Spritzen Spritzen, medizinisch
PICO syringes helps to deliver a blood sample with high analytical quality. Designed with uniquely formulated dry, electrolyte-balanced heparin that prevents clots from forming in the sample and minimises bias on all electrolytes. Includes standard luer tip, allows the syringes to be attached to the arterial catheder/line.
The syringe features TIPCAP that securely attached to the syringe and reduces the risk of blood spillage during transport. The standard TIPCAP is designed with air channels that prevent air from entering into the blood sample as the syringe is being sealed.