Anti-IgG Goat Antibody

Lieferant: MP Biomedicals
ICNA08670281EA 318 EUR
ICNA08670281
Anti-IgG Goat Antibody
Antikörper
This product is developed in goat using mouse IgG purified from pooled normal mouse serum as the immunogen. IgG is the main antibody type found in plasma and extracellular fluid and is expressed on B cell membrane. It regulates several functions such as complement activation and phagocytosis and hence plays a pivotal role in facilitating cytological immune responses. Anti-mouse IgG (whole molecule) antibody can be used in T cell proliferation assays. It can also be used in Ouchterlony Double Diffusion and immunoelectrophoresis. Goat anti- mouse IgG (whole molecule) antibody reacts specifically with mouse IgG subclasses IgG1, IgG2a, IgG2b and IgG3. The product has also shown no reactivity to human serum proteins.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR