Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))

Lieferant: MP Biomedicals
ICNA0855690EA 344 EUR
ICNA0855690
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Antikörper
This product is horseradish peroxidase (HRP)-conjugated goat affinity purified antibody to rabbit IgG Fc and buffer salts. The total protein is measured using the Biuret procedure with bovine albumin as standard. To ensure that
the product titer falls within the required range, antibody titer is standardized by microtiter plate ELISA with rabbit IgG. The
product is tested for purity and specificity at final concentration by immunoelectrophoresis. The antibody is goat IgG; no trace of
albumin is detected. It shows reactivity to rabbit Fc and IgG; cross-reactivity to other species may exist. Antibody activity to
F(ab')2, IgA, IgM, or non-immunoglobulin serum proteins is not present.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR