Anti-SRBC Rabbit Polyclonal Antibody

Lieferant: MP Biomedicals
ICNA0855806EA 316 EUR
ICNA0855806
Anti-SRBC Rabbit Polyclonal Antibody
Antikörper
Product is the lyophilized powder of rabbit IgG fraction to sheep red blood cells and buffer salts.

Type: Primary
Antigen: SRBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR