Anti-C3 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: MP Biomedicals
ICNA0855500EA 434 EUR
ICNA0855500
Anti-C3 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Product is the lyophilized powder of fluorescein-5-isothiocyanate (FITC "Isomer I")-conjugated goat IgG fraction to mouse complement C3 and buffer salts.

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR