Anti-Smad6 Rabbit Polyclonal Antibody [clone: N372]

Lieferant: Bioworld Technology
BWRLBS3555EA 262 EUR
BWRLBS3555
Anti-Smad6 Rabbit Polyclonal Antibody [clone: N372]
Antikörper
Synthetic peptide, corresponding to amino acids 341-390 of Human Smad6.

Smad6 (N372) pAb detects endogenous levels of Smad6 protein.

Type: Primary
Antigen: Smad6
Clonality: Polyclonal
Clone: N372
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR