Anti-OCT-1 Rabbit Polyclonal Antibody [clone: L68]

Lieferant: Bioworld Technology
BWRLBS1260EA 262 EUR
BWRLBS1260
Anti-OCT-1 Rabbit Polyclonal Antibody [clone: L68]
Antikörper
Synthetic peptide, corresponding to amino acids 30-80 of Human Oct-1.

OCT (L68) pAb detects endogenous levels of OCT1 protein.

Type: Primary
Antigen: OCT-1
Clonality: Polyclonal
Clone: L68
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR