Aluminiumoxid, Ecochrom™ Alumina A 63-200 µm Activity: I
Lieferant: MP Biomedicals
Synonyme:
Alumina C
ICNA0202105EA
284
EUR
ICNA0202105
Aluminiumoxid, Ecochrom™ Alumina A 63-200 µm Activity: I
Aluminium oxide
Specific Surface Area: 150 m2/g
Particle Size: 63-200 µm
Weight: 0.8 g/ml pH: 4.5
pH: 4.5
Formel:
Al₂O₃ Molecular Weight: 101,96 g/mol Dichte: 3,94 g/cm³ (20 °C) (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003424 CAS-Nummer: 1344-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...