Cleanroom garnment standard racks, CRG models
TINMPGH8S502000EA 699 EUR
TINMPGH8S502000 TINMPGH32S502000 TINMPGH16S502000 TINMPGH40S502000 TINMPGH40S501800 TINMPGH24S501800 TINMPGH8S501800 TINMPGH32S501800 TINMPGH24S502000 TINMPGH16S501800
Cleanroom garnment standard racks, CRG models
This garment racks design line and ensure ease of use and maintenance.
  • Rounded edges
  • Air permeable perforated steel frame for hangers
  • Stainless steel fixed hangers: clothing does not touch in between
  • Non-slip leveling feet
  • Minization of gaps: preventing accumulation of dust and for an easy cleaning
  • Simple assembly
Made of stainless steel AISI 304 with high 2R gloss polished finish
Cleanroom suitability: ISO class 4 - GMP class A
Lieferumfang: Detailed assembly instructions in every package