Cleanroom finger cots, latex
Lieferant: Basan
BSNC380015EA 0 EUR
BSNC380015 BSNC380018 BSNC380017 BSNC380016
Cleanroom finger cots, latex
Fingerlinge Fingerlinge, Latex
Conductive black finger cots made of latex.
  • Powder-free
  • Surface resistance: 10⁶/10⁸ Ω
  • Washed in an ISO 6 cleanroom
  • Packed in anti-static shielded bags
Recommended for use with electrostatic sensitive devices/components in ESD areas, production environments (e.g. electronics, jewellery) and cleanrooms (ISO class 5).
Colour: Black
Standards: EN 455