Immunoperoxidase systems, VECTASTAIN® Elite® ABC Kit (Standard)
Lieferant: Vector Laboratories
VECTPK-6100EA 941 EUR
VECTPK-6100
Immunoperoxidase systems, VECTASTAIN® Elite® ABC Kit (Standard)
Proteinkonjugate und Markierungen
The VECTASTAIN® Elite® ABC kit is supplied without biotinylated secondary antibodies or blocking serum. The 'Standard' kit consists of Elite® A and B reagents, which can be used with any biotinylated primary or secondary antibody, biotinylated nucleic acid probe or other biotinylated molecule.
Bestellinformation: Each kit contains sufficient reagents to prepare 110 ml of each working solution (500 to 1000 sections).