Laboratory freeze dryers, LSC
Lieferant: Martin Christ
LSC
535-3161EA 0 EUR
535-3161 535-3160 535-3159 535-3163 535-3162
Laboratory freeze dryers, LSC
Gefriertrockner
Universal freeze dryer for advanced processes; low ice condenser temperature allow drying of samples containing organic solvents (e.g. alcohols); with new intuitive understandable unit control LSC 'Lyo Screen Control':
  • ¹/₄ VGA-LC-Display
  • Graphic orientated display with single button control ('turn and push')
  • Clear structured display of relevant process data
  • Storage of up to 30 freeze drying programs in tables (option)
  • Ice condenser temperature: ∼−60 or −85 °C
Bestellinformation: WINDOWS based process control and documentation software is available.