Dual stage pressure regulators
Lieferant: Kayser-Werk
KAYSCK1310EA 368 EUR
KAYSCK1310
Dual stage pressure regulators
Druckminderer
The set outlet pressure is kept constant regardless of the bottle level.
Standards: Manufactured according to EN ISO 2503.