Single and dual stage gas regulators, Silcosteel®
Lieferant: Restek
REST21360-2EA 0 EUR
REST21360-2 REST21361-5
Single and dual stage gas regulators, Silcosteel®
Druckminderer
These single- and dual-stage regulators are available with Silcosteel® surface treatment. This proprietary passivation process provides excellent inertness for sulfur and mercury calibration standards and improved corrosion resistance over bare 316L stainless steel or other more expensive alloys.
  • Applicable to a number of varied purposes
  • Useful in gauging environmental stack and gas emission standards
  • Outlet pressure of 0 to 150 psig