Slide warmer/driers
Lieferant: Klinipath
KPXX12801EA 1560 EUR
KPXX12801 KPXX13801 KPXX15801
Slide warmer/driers
Objektträger-Trockner
These slide warmers have a digital display for showing actual temperature. Ideal for stretching and drying sectioned tissue samples in pathology, histology, pharmacology, biochemistry, biology, materials research, cosmetic industry, food industry and textile industries. The set temperature can be set up to 100 °C in increments of 1 °C and is digitally displayed. Electronic temperature regulation with an accuracy of < 1%.
  • Work surface covered with black epoxy giving good sample visibility
  • Electronic temperature control
  • Over-heating protection
Height: 70 mm.