Human prostate specific antigen (from Seminal fluid)
Lieferant: MP Biomedicals
ICNA08322601EA 322 EUR
ICNA08322601
Human prostate specific antigen (from Seminal fluid)
Enzyme
PSA supplied as solution in 0,05 M sodium phosphate buffer, pH 7,5, containing 0,15 M NaCl and 0,09% sodium azide. Sterile filtered using a 0,2 µm filter.
Donors are screened for Hepatitis B surface antigen (HBSAg), HIV 1/2, HCV antibodies.