Glass bead sterilisers
SAFSZ378585EU-1EAEA 0 EUR
SAFSZ378585EU-1EA
Glass bead sterilisers
This glass bead steriliser is primarily for sterilising plunge metal instruments which is kept into hot glass beads for instant sterilisation. The glass beads in the well are mantained at 250 °C for complete destruction of microorganisms and spores in just seconds.
  • Well and lid composition: rust resistant chrome-nickel steel
  • Rust resistant outer case
The glass beads can be cleaned by any cleaning liquid (or soapy water). Just rinse the glass beads thoroughly with hot water, dry completely and refill the steriliser. The chamber is deep enough for sterilising small and medium sized instruments.
Lieferumfang: Supplied with a two-pin EU plug.