Adenosine assay kit

Lieferant: Abnova
ABNOKA4547EA 958 EUR
ABNOKA4547
Adenosine assay kit
The adenosine assay kit is for the fluorometric determination of adenosine in plasma and urine.

  • Calibration range: 2 to 80 pmol
  • Reactivity: mammals
  • Storage temperature: −20 °C

Lieferumfang: The kit contains adenosine assay buffer (25 ml), urine clarifier (1 vial), adenosine detector (1 vial), adenosine convertor (1 vial), adenosine developer (1 vial), adenosine standard 10 mm (100 µl) and adenosine probe (DMSO) 200 µl.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR