Lactate assay kit
Lieferant: Abnova
ABNOKA0833EA 958 EUR
ABNOKA0833
Lactate assay kit
Tests Enzymtests
Lactate Assay Kit is used for detecting L(+)-Lactate. Suitable samples include biological fluids, cell culture supernatant, plasma, serum, tissue culture supernatant and urine.
Lactate specifically reacts with an enzyme mix to generate a product, which interacts with lactate probe to produce colour (OD 570 nm) and fluorescence (Ex/Em=535/587 nm). There is no need of pretreatment or purification of samples. The kit can detect 0,001 to 10 mM of lactate in various samples.
Lieferumfang: Kit includes Lactate Assay Buffer, 25 ml; Lactate Probe (in DMSO, anhydrous), 200 µl; Lactate Enzyme Mix (lyophilised), 1 vial; and L(+)-Lactate Standard (100 nmol/µl), 100 µl. A clear 96-well plate is required but not supplied.