Total organic carbon (TOC) vial
Lieferant: IONICS
IONICHMI90606EA 922 EUR
IONICHMI90606
Total organic carbon (TOC) vial
Vials
TOC vials are designed for use in critical pharmaceutical applications such as USP, EP, JP and KP water testing.