VWR®, Sampling Bottles, Wide Mouth, Square, PET, with Tamper Evident Screw Cap
Lieferant: LAB3 OWN PRODUCTION
VWRLPET250AEA 3000 EUR
VWRLPET250A VWRL080214
VWR®, Sampling Bottles, Wide Mouth, Square, PET, with Tamper Evident Screw Cap
Flaschen Kunststoffflaschen
Bottles, made of PET, amber with white PP cap. Ideal for water sampling and general sample collection.
  • Amber coloured for light sensitve samples
  • Lightweight and durable
  • Leakproof
  • Temperature resistance −10 to +40 °C
  • Not autoclavable
  • Screw caps must be ordered separately