Microplates, 1536-well, multi-coated, Corning®
Lieferant: Corning
Corning®
CORN3829EA 1613.96 EUR
CORN3829
Microplates, 1536-well, multi-coated, Corning®
Mikrotiterplatten HTS-Mikrotiterplatten
Multi-coated microplate, PS, flat bottom, black with clear base, low base design, with lid. Black wall plates have low background fluorescence, minimal light scatter and reduced crosstalk.
  • No logo or lettering for improved heat sealing and ease of liquid handling
  • Low base designed for optimal bottom reader sensitivity
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness minimises variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers
  • Six different surface treatments on a single plate (collagen I, fibronectin, gelatin, laminin, poly-D-lysine, TC-treated)