Microplates, 384-well, multi-coated, Corning®
Lieferant: Corning
Corning®
CORN4589EA 850.36 EUR
CORN4589
Microplates, 384-well, multi-coated, Corning®
Mikrotiterplatten HTS-Mikrotiterplatten
Multi-coated microplate, PS, black with clear, flat bottom, with lid. Black wall plates have low background fluorescence, minimal light scatter and reduced crosstalk.
  • Opaque walls to prevent well to well crosstalk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments
  • Individual alphanumeric codes for well identification