Wipe roll, MicroRoll CE
Lieferant: Vileda Professional
VILE149145EA 234 EUR
VILE149145
Wipe roll, MicroRoll CE
Disposable microfire cloth for cleanroom cleaning.
  • Perforated roll with 200 cloth (25×35 cm)
  • Streak free surface cleaning esp. on stainless steel, glass and mirrors
  • No linting, high tear resistance
Designed for use in cleanroom class ISO 7–9 / GMP C-D.