Cleaning trolley Clino®, CR4 FP-CR
Lieferant: Pfennig
PPSA3540200EA 2033.15 EUR
PPSA3540200
Cleaning trolley Clino®, CR4 FP-CR
The Clino® CR4 FP-CR trolley system made of stainless steel suitable for cleanrooms and for high sensitive areas that require minimal space. ClinoLink cleanroom-suitable connection system for individual attachment of accessory parts.
  • Individually configurable through Clino modular design system
  • Certified flat wringer ringo GMP for sterile areas
  • Full flexibility in single- and two-step technique
  • Basic module mini with high end finish and castors (100 mm)
  • Includes contact-free dropping device MopDrop
  • Wiper impregnation for surface cleaning in Clino PlusBox
It is suitable for technical cleanrooms ISO classes 7 and 8.
Lieferumfang: Supplied with 2 ×PlusBoxes, bucket (25 L and 15 L), disposal unit (1 ×35 L) and plastic container (5 L).