Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: MP Biomedicals
ICNA08674171EA 555 EUR
ICNA08674171
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Based on immunoelectrophoresis, these antibodies react with the heavy chains on human IgG but not with the light chains on most human immunoglobulins. No antibody was detected against human IgM or IgA or non-immunoglobulin serum proteins, but antibodies may cross-react with immunoglobulins from other species.

  • Working dilution: 1:500 - 1:5000 for immunochemistry on tissue sections
  • Working dilution: 1:5000 - 1:100 000 for ELISA and western blots with chromogenic substrates
  • Working dilution: 1:10 000 - 1:200 000 for western blot with ECL substrates

Type: Secondary
Antigen: IgG Fc
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horse radish peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR