Anti-Ig IgM Rabbit Polyclonal Antibody

Lieferant: MP Biomedicals
ICNA08643901EA 386 EUR
ICNA08643901
Anti-Ig IgM Rabbit Polyclonal Antibody
Antikörper
Based on Immunoelectrophoresis and Immunodiffusion, it produces one IgM band against pig serum and no bands against pig IgG.

  • Can be reconstitute with 2,0 ml of distilled water

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR