ICNA02150958

Lieferant: MP Biomedicals
ICNA0215095890EA 150 EUR
ICNA0215095890
ICNA02150958
Dimidium Bromide is used as fluorescent probe for nucleic acids and as interchelating agent.

  • Presentation: Dark Reddish Purple Powder
  • Soluble in water (4 mg/mL - clear to hazy orangish-red solution).
  • Storage temperature: Store at Room temperature (15 - 30 °C).
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR