Labels for BMP™ 71 printer, type B-427
Lieferant: Brady
116-0428EA 38.13 EUR
116-0428 BRDY114969 BRDY114804 BRDY114965 BRDY114964 BRDY114966 BRDY114960 BRDY114963 BRDY114820 BRDY115033 BRDY115099 BRDY115031 BRDY115032 BRDY115098 BRDY115095 BRDY115030 BRDY115093 BRDY115094 BRDY114818 BRDY114935 BRDY114932 BRDY114811 BRDY114877 BRDY114876 BRDY114931 BRDY114934 BRDY114879 BRDY114878 BRDY115108 BRDY114933 BRDY114950 BRDY114994 BRDY114873 BRDY114872 BRDY114971 BRDY115029 BRDY115103 BRDY114875 BRDY114874 BRDY114951 BRDY114990 BRDY115101 BRDY115102 BRDY114871 BRDY114970 BRDY114794 BRDY115089 BRDY115100
Labels for BMP™ 71 printer, type B-427
Etiketten
Self-laminating vinyl tape/labels are ideal for communications cables, laboratory and panel identification.
  • Good water and oil resistance
  • Good abrasion and smudge resistance
This material is UL approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.