Anti-S100A9426 Mouse Monoclonal Antibody [clone: CAGB/426]
Lieferant: Abnova
ABNOMAB13427EA 440 EUR
ABNOMAB13427
Anti-S100A9426 Mouse Monoclonal Antibody [clone: CAGB/426]
Antikörper
Mouse monoclonal antibody raised against full length recombinant human S100A9.
Type: Primary
Antigen: S100A9426
Clonality: Monoclonal
Clone: CAGB/426
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human