Anti-LARP1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB29903EA 518 EUR
ABNOPAB29903
Anti-LARP1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against partial synthetic protein of human LARP1.

Type: Primary
Antigen: LARP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR