Anti-CREB3L2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB30010EA 338 EUR
ABNOPAB30010
Anti-CREB3L2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of human CREB3L2.

Type: Primary
Antigen: CREB3L2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR