Hygromycin B 50 mg/ml in PBS
Lieferant: MIRUS
MIRUMIR5930EA 283 EUR
MIRUMIR5930
Hygromycin B 50 mg/ml in PBS
Hygromycin B
Formel: C₂₀H₃₇N₃O₁₃
Molecular Weight: 527,5 g/mol
MDL: MFCD06795479
CAS-Nummer: 31282-04-9