2-Bromanthracen 98%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO454880010EA
180
EUR
ACRO454880010
2-Bromanthracen 98%
2-Bromoanthracene
Formel:
C₁₄H₉Br Molecular Weight: 257,1300 g/mol Schmelzpunkt: 220…221,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784002 CAS-Nummer: 7321-27-9 |
Spezifikation Testergebnisse
Appearance | Yellow to light brown Powder |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...