6-Chinoxalincarbonsäurepinakolester 97%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO445520010EA
190
EUR
ACRO445520010
6-Chinoxalincarbonsäurepinakolester 97%
6-Chinoxalincarbonsäurepinakolester
Formel:
C₁₄H₁₇BN₂O₂ Molecular Weight: 256,1080 g/mol Siedepunkt: 130…145 °C (,6 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD11054040 CAS-Nummer: 1167418-13-4 |
Spezifikation Testergebnisse
Appearance | Yellow to light brown Viscous liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...