4-Cyan-2-fluorphenylboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO445390010EA
364
EUR
ACRO445390010
4-Cyan-2-fluorphenylboronsäure 97%
4-Cyan-2-fluorphenylboronsäure
Formel:
C₇H₅BFNO₂ Molecular Weight: 164,93 g/mol Schmelzpunkt: 304…304,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07644472 CAS-Nummer: 1150114-77-4 |
Spezifikation Testergebnisse
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...