3-Amino-4-brom-1H-indazol 95%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO435790050EA
213
EUR
ACRO435790050
3-Amino-4-brom-1H-indazol 95%
3-Amino-4-bromo-1H-indazole
Formel:
C₇H₆BrN₃ Molecular Weight: 212,05 Schmelzpunkt: 148…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09909580 CAS-Nummer: 914311-50-5 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Off-white to grey to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 147 to 152 °C |
HPLC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...