3-Cyan-5-fluorphenylboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO430460010EA
211
EUR
ACRO430460010
3-Cyan-5-fluorphenylboronsäure 97%
3-Cyan-5-fluorphenylboronsäure
Formel:
C₇H₅BFNO₂ Molecular Weight: 164,9300 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09263608 CAS-Nummer: 304858-67-1 |
Spezifikation Testergebnisse
Appearance | White Crystalline solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration Argentometric | 97.5 to 102.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...