Benzyl-4-bromphenylketon
Lieferant: Thermo Scientific
|
Achtung
|
ACRO381890050EA
139
EUR
ACRO381890050
Benzyl-4-bromphenylketon
Benzyl-4-bromphenylketon
Formel:
C₁₄H₁₁BrO Molecular Weight: 275,14 g/mol Schmelzpunkt: 109…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016331 CAS-Nummer: 2001-29-8 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 110 to 115 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...